Learn More
Medchemexpress LLC CD28 Rat HEK293 F 10ug
SDP
Catalog No. 5000178381
Encompass_Preferred
CD28 Protein activates T-cells promoting cell proliferation cytokine production and T-cell survival It collaborates with TCR/CD3 ligation and CD40L costimulation to boost IL4 and IL10 production in T-cells CD28 Protein forms a disulfide-linked homodimer and interacts with DUSP14 It also binds to CD80/B7-1 and CD86/B7-2/B70 and interacts with GRB2 (By similarity) CD28 Protein Rat (HEK293 Fc) is the recombinant rat-derived CD28 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-78381 Supplier Medchemexpress LLC Supplier No. HYP7431010UG
CD28 Protein activates T-cells promoting cell proliferation cytokine production and T-cell survival It collaborates with TCR/CD3 ligation and CD40L costimulation to boost IL4 and IL10 production in T-cells CD28 Protein forms a disulfide-linked homodimer and interacts with DUSP14 It also binds to CD80/B7-1 and CD86/B7-2/B70 and interacts with GRB2 (By similarity) CD28 Protein Rat (HEK293 Fc) is the recombinant rat-derived CD28 protein expressed by HEK293 with C-hFc labeled tag