Learn More
Genscript Corporation CD28H/IGPR-1 hFc Chimera Huma
Catalog No. 5000058306
Encompass
CD28H is constitutively expressed on all naive T cells Repetitive antigenic exposure however induces a complete loss of CD28H on many T cells and CD28H negative T cells have a phenotype of terminal differentiation and senescence After extensive screening in a receptor array a B7-like molecule B7 homologue 5 (B7-H5) was identified as a specific ligand for CD28H
Catalog No. 50-000-58306 Supplier Genscript Corporation Supplier No. Z04184100
CD28H is constitutively expressed on all naive T cells Repetitive antigenic exposure however induces a complete loss of CD28H on many T cells and CD28H negative T cells have a phenotype of terminal differentiation and senescence After extensive screening in a receptor array a B7-like molecule B7 homologue 5 (B7-H5) was identified as a specific ligand for CD28H