Learn More
ACROBiosystems Recombinant Protein;Mouse CD28 Protein;HEK293;100UG;CD8,CD28,Tp44
Catalog No. 501658747
Encompass_Preferred
Recombinant Protein;Mouse CD28 Protein;100UG;Mouse CD28, His Tag (CD8-M52H6) is expressed from human 293 cells (HEK293). It contains AA Asn 20 - Lys 149 (Accession # NP_031668.3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17 kDa. The protein migrates as 30-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8747 Supplier ACROBiosystems Supplier No. CD8M52H6100UG
Recombinant Protein;Mouse CD28 Protein;100UG;Mouse CD28, His Tag (CD8-M52H6) is expressed from human 293 cells (HEK293). It contains AA Asn 20 - Lys 149 (Accession # NP_031668.3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17 kDa. The protein migrates as 30-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.