Learn More
ACROBiosystems Recombinant Protein;Human / Cynomolgus / Rhesus macaque CD28 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;100UG;CD8,CD28,Tp44
Catalog No. 501658188
Encompass_Preferred
Recombinant Protein;Human / Cynomolgus / Rhesus macaque CD28 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Human / Cynomolgus / Rhesus macaque CD28, mouse IgG2a Fc tag (CD8-H52A5) is expressed from human 293 cells (HEK293). It contains AA Asn 19 - Pro 152 (Accession # P10747-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 42.0 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human / Cynomolgus / Rhesus macaque CD28, mouse IgG2a Fc tag (Cat. No. CD8-H52A5) at 5 μg/mL (100 μL/well) can bind Human B7-2, Fc Tag (Cat. No. CD6-H5257) with a linear range of 0.313-5 μg/mL (QC tested).;Immobilized Human / Cynomolgus / Rhesus macaque CD28, mouse IgG2a Fc tag (Cat. No. CD8-H52A5) at 5 μg/mL (100 μL/well) can bind Human B7-1, Fc Tag (Cat. No. B71-H5259) with a linear range of 0.625-10 μg/mL (Routinely tested).
Catalog No. 50-165-8188 Supplier ACROBiosystems Supplier No. CD8H52A5100UG
Recombinant Protein;Human / Cynomolgus / Rhesus macaque CD28 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Human / Cynomolgus / Rhesus macaque CD28, mouse IgG2a Fc tag (CD8-H52A5) is expressed from human 293 cells (HEK293). It contains AA Asn 19 - Pro 152 (Accession # P10747-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 42.0 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human / Cynomolgus / Rhesus macaque CD28, mouse IgG2a Fc tag (Cat. No. CD8-H52A5) at 5 μg/mL (100 μL/well) can bind Human B7-2, Fc Tag (Cat. No. CD6-H5257) with a linear range of 0.313-5 μg/mL (QC tested).;Immobilized Human / Cynomolgus / Rhesus macaque CD28, mouse IgG2a Fc tag (Cat. No. CD8-H52A5) at 5 μg/mL (100 μL/well) can bind Human B7-1, Fc Tag (Cat. No. B71-H5259) with a linear range of 0.625-10 μg/mL (Routinely tested).