Learn More
Biocare Medical, LLC CD3 Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC CME324A
Encompass_Preferred
This rabbit monoclonal antibody reacts with the intracytoplasmic portion of the CD3 antibody antigen expressed by T cells. Studies have shown that CD3 stains human T-cells in both the cortex and medulla of the thymus and in peripheral lymphoid tissues. It does not react with B-cells, monocytes, granulocytes and platelets. CD3 is regarded as a reliable pan T-cell antibody used in the immunophenotyping of T-cell lymphomas in paraffin sections with the majority of T-cell lymphomas expressing positivity for CD3. When used in conjunction, CD3 and UCHL-1 together identified the vast majority of T-cell lymphomas in paraffin sections.
Catalog No. 50-827-23