Learn More
BD Cell Analysis 3P CD3
Supplier:  BD Cell Analysis 3P 641406
Encompass
Concentration: 100ug/ml; volume per test: 5ul; Isotype: Mouse BALB/c IgG1; Reactivity: Human (QC Testing); Application: Flow cytometry routinely tested); Immunogen: Human Thymocytes; Storage Buffer: Phosphate buffered saline with BSA and ProClin. CD3 clone SK7 is derived from the hybridization of mouse NS-1 myeloma cells with spleen cells from BALB/c mice immunized with human thymocytes. The CD3 antibody reacts with the epsilon chain of the CD3 antigen/T-cell antigen receptor (TCR) complex, and is composed of at least 6 proteins that range in molecular weight from 20-30kDa. APC-H7 is an APC-cyanine tandem fluorochrome, which uses an analog of Cy7 and has similar spectral properties to APC-Cy7. APC-H7 conjugates provide greater stability in light and paraformaldehyde fixatives and have less spillover into the APC channel than APC-Cy7 conjugates. APC-Cy7 and APC-H7 cannot be used simultaneously.
Catalog No. 50-627-002
Please call customer service for current pricing. 1-800-766-7000