Learn More
Diagnostic Biosystems CD3 CD3 (SP7)
Catalog No. 502407479
Encompass_Preferred
The CD3 antigen is present on early thymocytes and mature T cells and is generally regarded as a pan-T cell marker. This antibody will help detect CD3 expression in normal and neoplastic tissues. This antibody reacts with the intracytoplasmic portion of the CD3 antigen expressed by T cells. It stains human T cellsin both the cortex and medulla of the thymus and in peripheral lymphoid tissues. This antibody issuitable for staining normal and neoplastic T cells in formalin-fixed, paraffin-embedded tissues.
Catalog No. 50-240-7479 Supplier Diagnostic Biosystems Supplier No. RMPD005
The CD3 antigen is present on early thymocytes and mature T cells and is generally regarded as a pan-T cell marker. This antibody will help detect CD3 expression in normal and neoplastic tissues. This antibody reacts with the intracytoplasmic portion of the CD3 antigen expressed by T cells. It stains human T cellsin both the cortex and medulla of the thymus and in peripheral lymphoid tissues. This antibody issuitable for staining normal and neoplastic T cells in formalin-fixed, paraffin-embedded tissues.