Learn More
Diagnostic Biosystems CD3 CD3, T Cell
Catalog No. 502407453
Encompass_Preferred
This antibody reacts with intracytoplasmic portion of the CD3 antigen on the T cells. It stains T cells in cortex as well as medulla of the thymus and lymphoid tissues. This antibody also labels T cell neoplasm, malignant histiocytes and Reed-Sternberg cells in Hodgkin disease.
Catalog No. 50-240-7453 Supplier Diagnostic Biosystems Supplier No. PDR002
This antibody reacts with intracytoplasmic portion of the CD3 antigen on the T cells. It stains T cells in cortex as well as medulla of the thymus and lymphoid tissues. This antibody also labels T cell neoplasm, malignant histiocytes and Reed-Sternberg cells in Hodgkin disease.