Learn More
CAPRICO BIOTECHNOLOGIES INC CD3 APC-IFLUOR 700, Caprico Biotechnologies, Catalog Number 4053175
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4053175
Encompass_Preferred
The CD3 APC-IFLUOR 700 antibody from Caprico Biotechnologies is a mouse monoclonal antibody that targets human CD3. It is conjugated to APC-IFLUOR 700 and is recommended for flow cytometry applications. This antibody recognizes the epsilon chain of the CD3 complex and can be used to identify T lymphocytes. According to the product details, this antibody is supplied at 0.5 mg/mL in PBS with 0.1% sodium azide and 1% BSA. The recommended dilution for flow cytometry is 5-25 μL per test. Overall, this APC-IFLUOR 700-conjugated anti-human CD3 antibody from Caprico Biotechnologies enables identification and analysis of T cells by flow cytometry.
Catalog No. 50-227-9101