Learn More
Biolegend Brilliant Violet 421™ anti-human CD3
Supplier:  Biolegend 300434
Encompass_Preferred
Brilliant Violet 421™ anti-human CD3 [UCHT1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC, ICC, IHC-F; Size: 100 tests
Catalog No. NC0245650