Learn More
Biolegend Brilliant Violet 605™ anti-mouse CD3ε
Supplier:  Biolegend 100351
Encompass_Preferred
Brilliant Violet 605™ anti-mouse CD3ε [145-2C11]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC1327818