Learn More
BD Cell Analysis 3P CD3 FITC / CD8 PE / CD45 PerCP / CD4 APC Reagent, 50 tests
Catalog No. 50620182
Encompass
BD Multitest CD3 fluorescein isothiocyanate (FITC)/CD8 phycoerythrin (PE)/CD45 peridinin chlorophyll protein (PerCP)/CD4 allophycocyanin (APC) is a four-color direct immunofluorescence reagent for identifying and determining the percentages and absolute counts of mature human T lymphocytes (CD3+), suppressor/cytotoxic (CD3+CD8+) T-lymphocyte subsets, and helper/inducer (CD3+CD4+) T-lymphocyte subsets in erythrocyte-lysed whole blood. When used with BD Trucount absolute count tubes, these populations can be enumerated from a single tube.
Catalog No. 50-620-182 Supplier BD Cell Analysis 3P Supplier No. 340499
BD Multitest CD3 fluorescein isothiocyanate (FITC)/CD8 phycoerythrin (PE)/CD45 peridinin chlorophyll protein (PerCP)/CD4 allophycocyanin (APC) is a four-color direct immunofluorescence reagent for identifying and determining the percentages and absolute counts of mature human T lymphocytes (CD3+), suppressor/cytotoxic (CD3+CD8+) T-lymphocyte subsets, and helper/inducer (CD3+CD4+) T-lymphocyte subsets in erythrocyte-lysed whole blood. When used with BD Trucount absolute count tubes, these populations can be enumerated from a single tube.