Learn More
Beckman Coulter CD3-ECD, UCHT1, 1 mL, ASR
Supplier:  Beckman Coulter IM2705U
Encompass_Preferred
CD3 is part of a bigger complex which includes the T Cell Receptor (TCR). This complex is expressed by mature T cells and by thymocytes. T cell activation may be induced when a foreign antigen is presented to the TCR through MHC complex.
Catalog No. NC0720544