Learn More
Medchemexpress LLC CD3 epsilon 1-27 pep 20ug | 20ug
SDP
Catalog No. 5000187805
Encompass_Preferred
CD3 epsilon is an important component of the TCR-CD3 complex on T lymphocytes and promotes TCR-mediated signaling When APC activates the TCR CD3 epsilon together with CD3D CD3G and CD3Z transmits signals across the cell membrane through ITAMs CD3 epsilon 1-27 peptide Protein Human (Biotinylated HEK293 hFc-Avi) is the recombinant human-derived CD3 epsilon 1-27 peptide protein expressed by HEK293 with C-Avi C-hFc labeled tag
Catalog No. 50-001-87805 Supplier Medchemexpress LLC Supplier No. HYP7808920UG
CD3 epsilon is an important component of the TCR-CD3 complex on T lymphocytes and promotes TCR-mediated signaling When APC activates the TCR CD3 epsilon together with CD3D CD3G and CD3Z transmits signals across the cell membrane through ITAMs CD3 epsilon 1-27 peptide Protein Human (Biotinylated HEK293 hFc-Avi) is the recombinant human-derived CD3 epsilon 1-27 peptide protein expressed by HEK293 with C-Avi C-hFc labeled tag