Learn More
Medchemexpress LLC CD3 epsilon Canine 10ug | 10ug
SDP
Catalog No. 5000187213
Encompass_Preferred
The CD3 protein on lymphocytes is a component of the TCR-CD3 complex and is critical for adaptive immune responses When APC is activated TCR signals transmitted by CD3D CD3E CD3G and CD3Z initiate pathways through ITAM CD3 epsilon Protein Canine (HEK293 Fc) is the recombinant canine-derived CD3 epsilon protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-87213 Supplier Medchemexpress LLC Supplier No. HYP7732010UG
The CD3 protein on lymphocytes is a component of the TCR-CD3 complex and is critical for adaptive immune responses When APC is activated TCR signals transmitted by CD3D CD3E CD3G and CD3Z initiate pathways through ITAM CD3 epsilon Protein Canine (HEK293 Fc) is the recombinant canine-derived CD3 epsilon protein expressed by HEK293 with C-hFc labeled tag