Learn More
Sino Biological Anti-CD3 epsilon/CD3e Antibody, Mouse Monoclonal
Supplier:  Sino Biological 90047MM12100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rhesus CD3 epsilon / CD3e ( Catalog#90047-C08H; XP_001097204.1; Met1-Asp117). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-222-3101