Learn More
Medchemexpress LLC CD3 epsilon Human 100ug
SDP
Catalog No. 5000184810
Encompass_Preferred
Recombinant human CD3 epsilon protein supplied as a lyophilized powder, expressed in HEK293 cells and carrying a C-terminal 6xHis tag. The protein is intended for T-cell signaling research, assay development, and biochemical characterization; it offers high purity and low endotoxin to support sensitive in vitro and cell-based assays.
  • Lyophilized powder for convenient storage and transport.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • C-terminal 6xHis tag facilitates purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg, suitable for sensitive assays.
  • Available in 10 μg, 50 μg, 100 μg, and 500 μg sizes.
  • Recommended reconstitution at ≥100 μg/mL; refer to lot-specific COA for buffer details.
Catalog No. 50-001-84810 Supplier Medchemexpress LLC Supplier No. HYP70506100UG
Add to Cart
Add to Cart
Recombinant human CD3 epsilon protein supplied as a lyophilized powder, expressed in HEK293 cells and carrying a C-terminal 6xHis tag. The protein is intended for T-cell signaling research, assay development, and biochemical characterization; it offers high purity and low endotoxin to support sensitive in vitro and cell-based assays.
  • Lyophilized powder for convenient storage and transport.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • C-terminal 6xHis tag facilitates purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg, suitable for sensitive assays.
  • Available in 10 μg, 50 μg, 100 μg, and 500 μg sizes.
  • Recommended reconstitution at ≥100 μg/mL; refer to lot-specific COA for buffer details.