Learn More
Medchemexpress LLC CD3 epsilon Human 500ug
SDP
Catalog No. 5000185332
Encompass_Preferred
Recombinant human CD3 epsilon protein produced in HEK293 mammalian cells and supplied as a lyophilized preparation with a C-terminal 6xHis tag, intended for research use in studies of T-cell receptor signaling, binding assays, and related immunological applications. Lot-specific COA provides detailed formulation and buffer information.
  • Produced in HEK293 mammalian expression system.
  • C-terminal 6xHis tag for detection and purification.
  • Lyophilized powder formulation for convenient storage and handling.
  • Approximate molecular weight 18-21 kDa by SDS-PAGE.
  • Stable at -20 °C for long-term storage; reconstituted stability at 4 °C for one week.
  • Lot-specific COA and datasheet available for quality and formulation details.
Catalog No. 50-001-85332 Supplier Medchemexpress LLC Supplier No. HYP70506500UG
Recombinant human CD3 epsilon protein produced in HEK293 mammalian cells and supplied as a lyophilized preparation with a C-terminal 6xHis tag, intended for research use in studies of T-cell receptor signaling, binding assays, and related immunological applications. Lot-specific COA provides detailed formulation and buffer information.
  • Produced in HEK293 mammalian expression system.
  • C-terminal 6xHis tag for detection and purification.
  • Lyophilized powder formulation for convenient storage and handling.
  • Approximate molecular weight 18-21 kDa by SDS-PAGE.
  • Stable at -20 °C for long-term storage; reconstituted stability at 4 °C for one week.
  • Lot-specific COA and datasheet available for quality and formulation details.