Learn More
Medchemexpress LLC CD3 epsilon Rat HE 5ug | 5ug
SDP
Catalog No. 5000185781
Encompass_Preferred
CD3 epsilon Protein a vital component of the TCR-CD3 complex on T-lymphocytes is pivotal for adaptive immune responses CD3E is crucial for proper T-cell development and contributes to TCR-CD3 complex internalization and down-regulation[1] CD3 epsilon Protein Rat (HEK293 Fc) is the recombinant rat-derived CD3 epsilon protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-85781 Supplier Medchemexpress LLC Supplier No. HYP767885UG
CD3 epsilon Protein a vital component of the TCR-CD3 complex on T-lymphocytes is pivotal for adaptive immune responses CD3E is crucial for proper T-cell development and contributes to TCR-CD3 complex internalization and down-regulation[1] CD3 epsilon Protein Rat (HEK293 Fc) is the recombinant rat-derived CD3 epsilon protein expressed by HEK293 with C-hFc labeled tag