Learn More
Medchemexpress LLC CD3 epsilon Rhesus 50ug | 50ug
SDP
Catalog No. 5000185785
Encompass_Preferred
CD3 epsilon Protein a vital component of the TCR-CD3 complex on T-lymphocytes is pivotal for adaptive immune responses CD3E is crucial for proper T-cell development and contributes to TCR-CD3 complex internalization and down-regulation[1] CD3 epsilon Protein Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived CD3 epsilon protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-85785 Supplier Medchemexpress LLC Supplier No. HYP7679150UG
CD3 epsilon Protein a vital component of the TCR-CD3 complex on T-lymphocytes is pivotal for adaptive immune responses CD3E is crucial for proper T-cell development and contributes to TCR-CD3 complex internalization and down-regulation[1] CD3 epsilon Protein Rhesus Macaque (HEK293 His) is the recombinant Rhesus Macaque-derived CD3 epsilon protein expressed by HEK293 with C-His labeled tag