Learn More
Sino Biological Human CD3e / CD3 epsilon Protein (His Tag) 500ug
Supplier:  Sino Biological 10977H08H500
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Asp 126) of human CD3E (NP_000724.1) precursor was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6444