Learn More
Sino Biological Recombinant Human CD3 epsilon/CD3e Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10977H02H50
Encompass_Preferred
A DNA sequence encoding the human CD3E (NP_000724.1) (Met1-Asp126) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-5049