Learn More
Enzo Life Sciences CD3 (human) monoclonal antibody (TR66) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804822C100
Encompass_Preferred
Clone: TR66. Isotype: IgG1. Immunogen: Cloned human helper T cell line. Source: Purified from concentrated hybridoma tissue culture supernatant. Species reactivity: Human. Specificity: Recognizes the ε-chain of CD3. Applications: Flow Cytometry, ICC, IP, FUNC. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9984