Learn More
CAPRICO BIOTECHNOLOGIES INC CD3 iFluor 488
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4053115
Encompass_Preferred
CD3 iFluor 488 is a monoclonal antibody that specifically binds to the CD3 antigen, a part of the T cell receptor complex expressed on the surface of T lymphocytes. This antibody is conjugated to the iFluor 488 fluorescent dye and is suitable for flow cytometry, immunofluorescence microscopy, and other fluorescence-based applications requiring the detection of CD3+ T cells. The antibody is available in multiple sizes and formats for flexibility. Overall, this reagent allows for specific labeling and analysis of T lymphocytes through binding of CD3.
Catalog No. 50-227-9102