Learn More
CAPRICO BIOTECHNOLOGIES INC CD3 iFluor 700 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4053195
Encompass_Preferred
The CD3 iFluor 700 Antibody is a mouse monoclonal antibody that specifically binds to human CD3 epsilon chain. It is conjugated to iFluor 700 and is suitable for flow cytometry, immunohistochemistry, and Western blot applications. This antibody recognizes the CD3 T cell co-receptor complex found on mature T lymphocytes and is useful for studying T cell development and activation. According to the product details, this antibody is verified for flow cytometry using human peripheral blood lymphocytes. Each vial contains 100 tests of antibody in PBS with 0.1% sodium azide and 0.2% BSA.
Catalog No. 50-227-9103