Learn More
Biocare Medical, LLC CD3 Antibody - Predilute, 6.0 ml
SDP
Supplier:  Biocare Medical, LLC PP215AA
Encompass_Preferred
CD3 antibody (P) reacts with the intracytoplasmic portion of the CD3 antigen expressed by T cells. Studies have shown that CD3 stains human T-cells in both the cortex and medulla of the thymus and in peripheral lymphoid tissues. CD3 is regarded as a reliable pan T-cell antibody used in the immunophenotyping of T-cell lymphomas in paraffin sections with the majority of T-cell lymphomas expressing positivity for CD3. Studies have shown that when used in conjunction with LCA and CD20 [L26], CD3 (P) can determine cell lineage in the majority of non-Hodgkin lymphoma.
Catalog No. 50-832-06