Learn More
Charles River Labs Cell Solutions CD3+ PAN T CELL;NEG SEL 25X106
Catalog No. NC1769365
Encompass
CD3+ Pan T Cells; Negative Selection 25 x 106 CRYOPRESERVEDSource: Peripheral BloodStatus: HealthyContains: CryoStor® CS10Purity: >90% by flow cytometryViability: >95% by flow cytometry
Catalog No. NC1769365 Supplier Charles River Labs Cell Solutions Supplier No. PB03NC3
CD3+ Pan T Cells; Negative Selection 25 x 106 CRYOPRESERVEDSource: Peripheral BloodStatus: HealthyContains: CryoStor® CS10Purity: >90% by flow cytometryViability: >95% by flow cytometry