Learn More
Biogenex Laboratories CD3 : PS1 : 6mL
SDP
Supplier:  Biogenex Laboratories AM3225M
Encompass_Preferred
Human CD3 complex is associated with the T cell receptor (TcR) at the cell surface. Expression of CD3 antigen is generally restricted to the T-cell lineage, but weak expression might also occur in Purkinje cells in the brain, in macrophages, and in Reed-Sternberg cells in Hodgkin's lymphoma. The CD3 antigen is expressed early in the maturation of T cells. Monoclonal antibody PS1 reacts with the non glycosylated epsilon chain of CD3.
Catalog No. 50-197-8759