Learn More
STEMCELL Technologies Anti-Human CD3 Antibody, Clone UCHT1, FITC, Size: 500 Tests
SDP
Supplier:  STEMCELL Technologies 1000447
Encompass
Clone UCHT1 is a mouse (BALB/c) monoclonal antibody that reacts with the ~20 kDa CD3ε subunit of the human and chimpanzee T cell receptor (TCR)/CD3 complex which is expressed on the surface of ~95% of mature T cells and NKT cells and variably on thymocytes. Verified for use with flow cytometry and cell separation assays use this antibody in applications requiring the detection of CD3. This antibody clone has also been verified for purity assessments of cells isolated with EasySep kits including EasySep Direct Human T Cell Isolation Kit (Catalog 19661) EasySep Human CD3 Positive Selection Kit II (Catalog 17851 partial blocking may be observed) EasySep HLA Whole Blood T Cell Enrichment Kit (Catalog 19951HLA) and EasySep HLA Whole Blood CD2 Positive Selection Kit (Catalog 18687HLA). Visit STEMCELL.com for more information and related products.
Catalog No. 50-239-0170