Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology CD3 Zeta Polyclonal Antibody, Rabbit Host, Human and Mouse Reactivity, 100 μL
SDP

Catalog No. 502730446
Encompass

This polyclonal antibody is designed to recognize endogenous levels of total CD3ζ protein. It is produced by immunizing animals with a synthetic peptide corresponding to residues near the carboxy terminus of the human CD3ζ protein, and purified via peptide affinity chromatography. The antibody is validated for western blotting applications.

  • Validated reactivity: human and mouse
  • Host species: rabbit
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA and 50% glycerol
  • Store at -20°C
  • Recommended dilution for western blotting: 1:1,000

Catalog No. 50-273-0446 Supplier Cell Signaling Technology Supplier No. 88083S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This polyclonal antibody is designed to recognize endogenous levels of total CD3ζ protein. It is produced by immunizing animals with a synthetic peptide corresponding to residues near the carboxy terminus of the human CD3ζ protein, and purified via peptide affinity chromatography. The antibody is validated for western blotting applications.

  • Validated reactivity: human and mouse
  • Host species: rabbit
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA and 50% glycerol
  • Store at -20°C
  • Recommended dilution for western blotting: 1:1,000

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.