Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 943MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: Ki-1/779. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Hodgkin.’s lymphoma. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human TNFRSF8 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 105-120kDa.Chromosome: 1p36.22. Gene Symbol: TNFRSF8. Gene ID: 943. Ensemble: ENSG00000120949. Uniprot: XP_011540745.
Catalog No. 50-188-5341