Learn More
CAPRICO BIOTECHNOLOGIES INC CD30 FITC Antibody, 4221015
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4221015
Encompass_Preferred
The Caprico CD30 FITC Antibody, a monoclonal antibody conjugated with FITC, targets CD30 on activated T/B lymphocytes, marking Hodgkin's lymphoma. Used in flow cytometry and immunostaining, it comes as 100 tests with a 12-month shelf life. Optimal dilution is user-determined, and storage is at 2-8°C.
Catalog No. 50-227-9113