Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD30 / TNFRSF8 Protein, Avitag™,His Tag
Catalog No. 501957430
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CD30, Avitag,His Tag (CD0-H82E6) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 41.1 kDa. The protein migrates as 60-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525b) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD30, Avitag,His Tag (Cat. No. CD0-H82E6) with a linear range of 10-78 ng/mL (QC tested).;Immobilized Anti-CD30 MAb, Human IgG1 at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD30, Avitag,His Tag (Cat. No. CD0-H82E6) with a linear range of 0.4-6.5 ng/mL (Routinely tested).
Catalog No. 50-195-7430 Supplier ACROBiosystems Supplier No. CD0H82E6200UG
Recombinant Protein;200UG;Biotinylated Human CD30, Avitag,His Tag (CD0-H82E6) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 41.1 kDa. The protein migrates as 60-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525b) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD30, Avitag,His Tag (Cat. No. CD0-H82E6) with a linear range of 10-78 ng/mL (QC tested).;Immobilized Anti-CD30 MAb, Human IgG1 at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD30, Avitag,His Tag (Cat. No. CD0-H82E6) with a linear range of 0.4-6.5 ng/mL (Routinely tested).