Learn More
Genscript Corporation CD30 Ligand/TNFSF8 mFc Chimera
Catalog No. 5000057228
Encompass
CD30 ligand (CD30L)/TNFSF8 is a type II membrane protein belonging to the TNF superfamily CD30L is expressed on the cell surface of activated T cells B cells and monocytes The protein is also constitutively expressed on granulocytes and medullary thymic epithelial cells CD30L is a cytokine that binds to TNFRSF8/CD30 Induces proliferation of T-cells
Catalog No. 50-000-57228 Supplier Genscript Corporation Supplier No. Z04186500
CD30 ligand (CD30L)/TNFSF8 is a type II membrane protein belonging to the TNF superfamily CD30L is expressed on the cell surface of activated T cells B cells and monocytes The protein is also constitutively expressed on granulocytes and medullary thymic epithelial cells CD30L is a cytokine that binds to TNFRSF8/CD30 Induces proliferation of T-cells