Learn More
Enzo Life Sciences CD30 Recombinant monoclonal antibody (Ki-1/1505R) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS3540100
Encompass_Preferred
Clone: Ki-1/1505R. Isotype: IgGκ. Immunogen: Partial human CD30. Species reactivity: Human. Applications: Flow Cytometry, IF, IHC (PS). Purity: Protein A affinity purified. Formulation: Liquid. In 1X PBS containing 0.1 mg/ml BSA (US sourced) and 0.05% sodium azide.
Catalog No. 50-201-2883