Learn More
Enzo Life Sciences CD30 Recombinant monoclonal antibody (Ki-3) (Mouse IgG2b kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4320200
Encompass_Preferred
Clone: Ki-3. Isotype: IgG2bκ. Immunogen: Human CD30. Species reactivity: Human. Applications: ELISA, Flow Cytometry, IF, IHC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2959