Learn More
Enzo Life Sciences CD30 Recombinant monoclonal antibody (Ki-4) (Mouse IgG1 kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4340200
Encompass_Preferred
Clone: Ki-4. Isotype: IgG1κ. Immunogen: Human CD30. Species reactivity: Human. Applications: ELISA, Flow Cytometry, IF, IHC, FUNC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2961