Learn More
Genscript Corporation CD30/TNFRSF8 His Cynomolgus
Catalog No. 5000058832
Encompass
The transmembrane receptor CD30 (TNFRSF8) and its ligand CD30L (CD153 TNFSF8) are members of the tumor necrosis factor (TNF) superfamily and display restricted expression in subpopulations of activated T-and B-cells in nonpathologic conditions CD30 expression is upregulated in various hematological malignancies including Reed-Sternberg cells in Hodgkin s disease (HD) anaplastic large cell lymphoma (ALCL) and subsets of Non-Hodgkin s lymphomas (NHLs)
Catalog No. 50-000-58832 Supplier Genscript Corporation Supplier No. Z04114100
The transmembrane receptor CD30 (TNFRSF8) and its ligand CD30L (CD153 TNFSF8) are members of the tumor necrosis factor (TNF) superfamily and display restricted expression in subpopulations of activated T-and B-cells in nonpathologic conditions CD30 expression is upregulated in various hematological malignancies including Reed-Sternberg cells in Hodgkin s disease (HD) anaplastic large cell lymphoma (ALCL) and subsets of Non-Hodgkin s lymphomas (NHLs)