Learn More
Genscript Corporation CD30/TNFRSF8Biotin His Av
Catalog No. 5000059512
Encompass
The transmembrane receptor CD30 (TNFRSF8) and its ligand CD30L (CD153 TNFSF8) are members of the tumor necrosis factor (TNF) superfamily and display restricted expression in subpopulations of activated T-and B-cells in nonpathologic conditions CD30 expression is upregulated in various hematological malignancies including Reed-Sternberg cells in Hodgkin s disease (HD) anaplastic large cell lymphoma (ALCL) and subsets of Non-Hodgkin s lymphomas (NHLs)
Catalog No. 50-000-59512 Supplier Genscript Corporation Supplier No. Z04063100
The transmembrane receptor CD30 (TNFRSF8) and its ligand CD30L (CD153 TNFSF8) are members of the tumor necrosis factor (TNF) superfamily and display restricted expression in subpopulations of activated T-and B-cells in nonpathologic conditions CD30 expression is upregulated in various hematological malignancies including Reed-Sternberg cells in Hodgkin s disease (HD) anaplastic large cell lymphoma (ALCL) and subsets of Non-Hodgkin s lymphomas (NHLs)