Learn More
ACROBiosystems ACROBiosystems Human CD300c / LMIR2 Protein, His Tag
Catalog No. 501957499
Encompass_Preferred
Recombinant Protein;100UG;Human CD300c, His Tag (CDC-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Arg 183 (Accession # AAH22279).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.7 kDa. The protein migrates as 25-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7499 Supplier ACROBiosystems Supplier No. CDCH5224100UG
Recombinant Protein;100UG;Human CD300c, His Tag (CDC-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Arg 183 (Accession # AAH22279).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.7 kDa. The protein migrates as 25-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.