Learn More
Genscript Corporation CD300LF hFc Chimera Human
Catalog No. 5000059026
Encompass
Murine norovirus (MNoV) is an important model of human norovirus (HNoV) and mucosal virus infection more broadly CD300lf as a proteinaceous receptor for MNoV Interestingly its paralogue CD300ld was also sufficient for MNoV infection in vitro CD300lf is essential for both oral and parenteral MNoV infection and to elicit anti-MNoV humoral responses in vivo human CD300lf (huCD300lf) is not essential for HNoV infection nor does huCD300lf inhibit binding of HNoV virus-like particles to glycans
Catalog No. 50-000-59026 Supplier Genscript Corporation Supplier No. Z04192500
Murine norovirus (MNoV) is an important model of human norovirus (HNoV) and mucosal virus infection more broadly CD300lf as a proteinaceous receptor for MNoV Interestingly its paralogue CD300ld was also sufficient for MNoV infection in vitro CD300lf is essential for both oral and parenteral MNoV infection and to elicit anti-MNoV humoral responses in vivo human CD300lf (huCD300lf) is not essential for HNoV infection nor does huCD300lf inhibit binding of HNoV virus-like particles to glycans