Learn More
ACROBiosystems Recombinant Protein;Human CD300LG / Nepmucin Protein;HEK293;1MG;CDG,CD300LG,CLM9,TREM4,CD300g,Nepmucin,CLM-9,TREM-4
Supplier:  ACROBiosystems CDGH52231MG
Encompass_Preferred
Recombinant Protein;Human CD300LG / Nepmucin Protein;1MG;Human CD300LG, His Tag (CDG-H5223) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 247 (Accession # AAH25395).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.6 kDa. The protein migrates as 33-68 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Catalog No. 50-165-8943