Learn More
ABclonal Technology CD301/CLEC10A Rabbit pAb
SDP
Supplier:  ABclonal Technology A1185920UL
Encompass_Preferred
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may function as a cell surface antigen. Two transcript variants encoding distinct isoforms have been identified for this gene.
Catalog No. 50-000-98895