Learn More
Biolegend Brilliant Violet 785™ anti-human CD303 (BDCA-2)
Supplier:  Biolegend 354222
Encompass_Preferred
Brilliant Violet 785™ anti-human CD303 (BDCA-2) [201A]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1106447