Learn More
ACROBiosystems Recombinant Protein;Human CD30 / TNFRSF8 Protein, Fc Tag;HEK293;100UG;CD0,TNFRSF8,CD30,TNFRSF-8,D1S166E,Ki-1
Catalog No. 501658945
Encompass_Preferred
Recombinant Protein;Human CD30 / TNFRSF8 Protein, Fc Tag;100UG;Human CD30, Fc Tag (CD0-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.6 kDa. The protein migrates as 100-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525b) at 5 μg/mL (100 μL/well) can bind Human CD30, Fc Tag (Cat. No. CD0-H5250) with a linear range of 2-20 ng/mL (QC tested).
Catalog No. 50-165-8945 Supplier ACROBiosystems Supplier No. CD0H5250100UG
Recombinant Protein;Human CD30 / TNFRSF8 Protein, Fc Tag;100UG;Human CD30, Fc Tag (CD0-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.6 kDa. The protein migrates as 100-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525b) at 5 μg/mL (100 μL/well) can bind Human CD30, Fc Tag (Cat. No. CD0-H5250) with a linear range of 2-20 ng/mL (QC tested).