Learn More
ACROBiosystems ACROBiosystems Human CD30 Ligand / TNFSF8 Protein, His Tag, low endotoxin
Supplier:  ACROBiosystems CDLH524B1MG
Encompass_Preferred
Recombinant Protein;1MG;Human CD30 Ligand, His Tag, low endotoxin (CDL-H524b) is expressed from human 293 cells (HEK293). It contains AA Gln 63 - Asp 234 (Accession # P32971-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 21.5 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD30, Fc Tag (Cat. No. CD0-H5250) at 2 μg/mL (100 μL/well) can bind Human CD30 Ligand, His Tag, low endotoxin (Cat. No. CDL-H524b) with a linear range of 0.039-0.313 μg/mL (QC tested).;Immobilized Human CD30 Ligand, His Tag, low endotoxin (Cat. No. CDL-H524b) at 2 μg/mL (100 μL/well) can bind Human CD30, Llama IgG2b Fc Tag, low endotoxin (Cat. No. TN8-H5250) with a linear range of 0.6-10 ng/mL (Routinely tested).
Catalog No. 50-195-6811