Learn More
ACROBiosystems Recombinant Protein;Human CD30 Ligand / TNFSF8 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;1MG;CDL,CD153 antigen,CD153,CD30 antigen ligand,CD30 Ligand,CD30L,CD30-L,CD30LCD30 ligand,CD30LGMGC138144,TNFSF8,tumor necrosis factor ligand superfamily member 8,CD30 ligand
Supplier:  ACROBiosystems CDLH525B1MG
Encompass_Preferred
Recombinant Protein;Human CD30 Ligand / TNFSF8 Protein, Mouse IgG2a Fc Tag, low endotoxin;1MG;Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (CDL-H525b) is expressed from human 293 cells (HEK293). It contains AA Gln 63 - Asp 234 (Accession # P32971-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 46.5 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD30, His Tag (Cat. No. CD0-H5229) at 2 μg/mL (100 μL/well) can bind Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525b) with a linear range of 0.4-6.25 ng/mL (QC tested).
Catalog No. 50-165-7808