Learn More
ACROBiosystems Recombinant Protein;Human CD30 / TNFRSF8 Protein, Llama IgG2b Fc Tag, low endotoxin;HEK293;1MG;TN8,TNFRSF8,CD30,TNFRSF-8,D1S166E,Ki-1
Catalog No. 501657903
Encompass_Preferred
Recombinant Protein;Human CD30 / TNFRSF8 Protein, Llama IgG2b Fc Tag, low endotoxin;1MG;Human CD30, Llama IgG2b Fc Tag, low endotoxin (TN8-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 66.3 kDa. The protein migrates as 95-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD30, Llama IgG2b Fc Tag, low endotoxin (Cat. No. TN8-H5250) at 0.5 μg/mL (100 μL/well) can bind Anti-CD30 Mab, Human IgG1 with a linear range of 0.1-1.6 ng/mL (Routinely tested).;Immobilized Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525b) at 0.5 μg/mL (100 μL/well) can bind Human CD30, Llama IgG2b Fc Tag, low endotoxin (Cat. No. TN8-H5250) with a linear range of 10-78 ng/mL (QC tested).
Catalog No. 50-165-7903 Supplier ACROBiosystems Supplier No. TN8H52501MG
Recombinant Protein;Human CD30 / TNFRSF8 Protein, Llama IgG2b Fc Tag, low endotoxin;1MG;Human CD30, Llama IgG2b Fc Tag, low endotoxin (TN8-H5250) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 66.3 kDa. The protein migrates as 95-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD30, Llama IgG2b Fc Tag, low endotoxin (Cat. No. TN8-H5250) at 0.5 μg/mL (100 μL/well) can bind Anti-CD30 Mab, Human IgG1 with a linear range of 0.1-1.6 ng/mL (Routinely tested).;Immobilized Human CD30 Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDL-H525b) at 0.5 μg/mL (100 μL/well) can bind Human CD30, Llama IgG2b Fc Tag, low endotoxin (Cat. No. TN8-H5250) with a linear range of 10-78 ng/mL (QC tested).