Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5175MSM7P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: C31.10. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Jurkat cells. Tonsil or Angiosarcoma. Cellular Staining: Cell surface, cytoplasm of endothelial cells. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human CD31 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: ~100kDa (endothelium), ~130kDa (platelets) .Chromosome: 17q23.3. Gene Symbol: PECAM1. Gene ID: 5175. Ensemble: ENSG00000261371. Uniprot: XP_005276940.
Catalog No. 50-188-5048