Learn More
ACROBiosystems ACROBiosystems Human CD31 / PECAM-1 Protein, Fc Tag
Catalog No. 501957497
Encompass_Preferred
Recombinant Protein;200UG;Human CD31, Fc Tag (CD1-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Lys 601 (Accession # AAH22512.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 90.6 kDa. The protein migrates as 100-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7497 Supplier ACROBiosystems Supplier No. CD1H5258200UG
Recombinant Protein;200UG;Human CD31, Fc Tag (CD1-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Lys 601 (Accession # AAH22512.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 90.6 kDa. The protein migrates as 100-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.